Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTACGTCTGCTGCGGAGGGCGGCG[C/T]GCGGAGCTGCGGCGGCGGCGCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLYBL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLYBL - citrate lyase beta like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206808.3 | 63 | Missense Mutation | CGC,TGC | R,C 11 | NP_996531.1 | |
XM_005254030.2 | 63 | Missense Mutation | CGC,TGC | R,C 11 | XP_005254087.1 | |
XM_005254031.2 | 63 | Missense Mutation | CGC,TGC | R,C 11 | XP_005254088.1 | |
XM_006719915.3 | 63 | Missense Mutation | CGC,TGC | R,C 11 | XP_006719978.1 | |
XM_011521051.2 | 63 | Missense Mutation | CGC,TGC | R,C 11 | XP_011519353.1 | |
XM_011521052.2 | 63 | Missense Mutation | CGC,TGC | R,C 11 | XP_011519354.1 | |
XM_017020402.1 | 63 | Missense Mutation | CGC,TGC | R,C 11 | XP_016875891.1 | |
XM_017020403.1 | 63 | UTR 5 | XP_016875892.1 | |||
XM_017020404.1 | 63 | Intron | XP_016875893.1 |