Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGGGGTATTTGTACTTCCCGC[C/T]TTCCCAGGCGTTTTCTTTTTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNMAL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNMAL2 - paraneoplastic Ma antigen family-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020709.2 | 1867 | Missense Mutation | AGC,GGC | S,G 474 | NP_065760.1 |
PPP5D1 - PPP5 tetratricopeptide repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205281.1 | 1867 | Intron | NP_001192210.1 | |||
XM_005258397.3 | 1867 | Intron | XP_005258454.1 | |||
XM_006722960.3 | 1867 | Intron | XP_006723023.1 | |||
XM_017026125.1 | 1867 | Intron | XP_016881614.1 |