Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTATTTGTGCAGAGATCTGCAT[A/G]TGTTCCTTCAATTCCTCTCTTCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604443 | ||||||||||||||||||||
Literature Links: |
ACSL6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSL6 - acyl-CoA synthetase long-chain family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009185.2 | 1807 | Missense Mutation | CAT,TAT | H,Y 646 | NP_001009185.1 | |
NM_001205247.1 | 1807 | Missense Mutation | CAT,TAT | H,Y 611 | NP_001192176.1 | |
NM_001205248.1 | 1807 | Missense Mutation | CAT,TAT | H,Y 621 | NP_001192177.1 | |
NM_001205250.1 | 1807 | Missense Mutation | CAT,TAT | H,Y 632 | NP_001192179.1 | |
NM_001205251.1 | 1807 | Missense Mutation | CAT,TAT | H,Y 546 | NP_001192180.1 | |
NM_015256.3 | 1807 | Missense Mutation | CAT,TAT | H,Y 646 | NP_056071.2 |