Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGACCATGTTTTCTGCACTTAC[A/G]AGTCTCTCCGCTGTGAACTTTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF251 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF251 - zinc finger protein 251 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138367.1 | 1802 | Missense Mutation | CGT,TGT | R,C 516 | NP_612376.1 | |
XM_005272356.4 | 1802 | Intron | XP_005272413.1 | |||
XM_006716683.2 | 1802 | Intron | XP_006716746.1 | |||
XM_011517361.1 | 1802 | Intron | XP_011515663.1 |