Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGATTATATTTTATTTCAGACG[C/G]AGGCATCGATCAAGCAGTAGCTCTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613352 | |||||||||||||||||||||||
Literature Links: |
RSRC1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RSRC1 - arginine and serine rich coiled-coil 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271834.1 | 372 | Silent Mutation | CGC,CGG | R,R 66 | NP_001258763.1 | |
NM_001271838.1 | 372 | Silent Mutation | CGC,CGG | R,R 66 | NP_001258767.1 | |
NM_016625.3 | 372 | Silent Mutation | CGC,CGG | R,R 66 | NP_057709.2 |