Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCATGCGAGAGGCACCTTCAGCT[C/T]GGCAGCGTGCTGCTCCCGACACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300408 MIM: 300281 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIPAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIPAP1 - GRIP1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020137.4 | 1751 | Missense Mutation | AAG,GAG | K,E 557 | NP_064522.3 | |
XM_011543935.1 | 1751 | Missense Mutation | AAG,GAG | K,E 567 | XP_011542237.1 | |
XM_011543936.1 | 1751 | Intron | XP_011542238.1 | |||
XM_011543937.1 | 1751 | Missense Mutation | AAG,GAG | K,E 491 | XP_011542239.1 | |
XM_017029656.1 | 1751 | Intron | XP_016885145.1 | |||
XM_017029657.1 | 1751 | Missense Mutation | AAG,GAG | K,E 491 | XP_016885146.1 | |
XM_017029658.1 | 1751 | Missense Mutation | AAG,GAG | K,E 481 | XP_016885147.1 | |
XM_017029659.1 | 1751 | Missense Mutation | AAG,GAG | K,E 481 | XP_016885148.1 | |
XM_017029660.1 | 1751 | Intron | XP_016885149.1 | |||
XM_017029661.1 | 1751 | Intron | XP_016885150.1 |
KCND1 - potassium voltage-gated channel subfamily D member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |