Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCCAGGATTGCTTCAACATTG[A/G]GCACCATTACTAGCTCCCCACCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611532 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6851 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6851 - microRNA 6851 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOL6 - nucleolar protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022917.4 | 3451 | Missense Mutation | CCC,CTC | P,L 1117 | NP_075068.2 | |
NM_139235.3 | 3451 | Missense Mutation | CCC,CTC | P,L 670 | NP_631981.2 | |
XM_017015044.1 | 3451 | Missense Mutation | CCC,CTC | P,L 510 | XP_016870533.1 |