Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGACAGAGGGAACCCCATCTAC[A/C]TGGGCCTGAATGGACTCAATCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605509 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL36A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL36A - interleukin 36, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014440.2 | 604 | Missense Mutation | ATG,CTG | M,L 65 | NP_055255.1 | |
XM_005263639.2 | 604 | Missense Mutation | ATG,CTG | M,L 65 | XP_005263696.1 | |
XM_011510965.1 | 604 | Missense Mutation | ATG,CTG | M,L 65 | XP_011509267.1 | |
XM_017003806.1 | 604 | Missense Mutation | ATG,CTG | M,L 65 | XP_016859295.1 |