Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAGATATTGAATCATGTTACCA[A/T]TTCTGTTTTTTTCCACCCTGTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605243 MIM: 612418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LY96 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LY96 - lymphocyte antigen 96 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195797.1 | 124 | Missense Mutation | ATT,TTT | I,F 4 | NP_001182726.1 | |
NM_015364.4 | 124 | Missense Mutation | ATT,TTT | I,F 4 | NP_056179.3 | |
XM_011517508.2 | 124 | Missense Mutation | ATT,TTT | I,F 4 | XP_011515810.1 | |
XM_017013299.1 | 124 | Missense Mutation | ATT,TTT | I,F 4 | XP_016868788.1 | |
XM_017013300.1 | 124 | Missense Mutation | ATT,TTT | I,F 4 | XP_016868789.1 |
TMEM70 - transmembrane protein 70 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |