Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGATCAGCAGGAGGAAGGCCAG[C/T]GCCAGGGTGAGGGCTAGCCAGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607985 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAGPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAGPA - N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016256.3 | 1395 | Silent Mutation | GCA,GCG | A,A 456 | NP_057340.2 | |
XM_011522517.2 | 1395 | Intron | XP_011520819.1 |
NAGPA-AS1 - NAGPA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC14L5 - SEC14 like lipid binding 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |