Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGGGCCCTGGACCTACTCCGGG[G/T]CCTGCCGCGTGTGAGCCTGGCCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611828 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL15 - mitochondrial ribosomal protein L15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014175.3 | 127 | Missense Mutation | GGC,GTC | G,V 18 | NP_054894.1 | |
XM_017013362.1 | 127 | Missense Mutation | GGC,GTC | G,V 18 | XP_016868851.1 |