Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCTATGCCCTGCAACTGCACC[A/C]GTTACAGGTAACCCGCGGGGGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP27A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP27A1 - cytochrome P450 family 27 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000784.3 | 682 | Missense Mutation | CAG,CCG | Q,P 83 | NP_000775.1 | |
XM_017003488.1 | 682 | Missense Mutation | AGT,CGT | S,R 7 | XP_016858977.1 |