Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTTCGCTGTGTTCGTCGCCGC[C/T]GCCGCCGCCACCGCCGTTGTCGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606479 MIM: 615116 MIM: 613936 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf74 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf74 - chromosome 17 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175734.4 | 455 | Missense Mutation | CGC,TGC | R,C 128 | NP_783861.3 | |
XM_006721471.3 | 455 | Missense Mutation | CGC,TGC | R,C 31 | XP_006721534.1 | |
XM_011523722.2 | 455 | UTR 5 | XP_011522024.1 |
LOC107983988 - proteoglycan 4-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NLGN2 - neuroligin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPEM1 - spermatid maturation 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM102 - transmembrane protein 102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |