Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGAATCCAGGTATATTGTGAACC[A/G]TAGTTTTGTTCAGTAATGAAGATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612517 MIM: 602616 MIM: 180469 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAAF2 - dynein axonemal assembly factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083908.1 | 2462 | Missense Mutation | ACG,ATG | T,M 746 | NP_001077377.1 | |
NM_018139.2 | 2462 | Missense Mutation | ACG,ATG | T,M 794 | NP_060609.2 |
MGAT2 - mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL36AL - ribosomal protein L36a like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |