Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGCATGTCTTCGGCCGGGGGGT[C/T]GCTGGTGGCGGCTGGGGGTGGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612081 MIM: 616951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL34 - interleukin 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTSS1L - metastasis suppressor 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138383.2 | 2622 | Missense Mutation | AAC,GAC | N,D 717 | NP_612392.1 | |
XM_005256236.4 | 2622 | Missense Mutation | AAC,GAC | N,D 815 | XP_005256293.2 | |
XM_011523450.2 | 2622 | Missense Mutation | AAC,GAC | N,D 761 | XP_011521752.1 | |
XM_011523453.2 | 2622 | Missense Mutation | AAC,GAC | N,D 869 | XP_011521755.2 | |
XM_011523454.2 | 2622 | Missense Mutation | AAC,GAC | N,D 866 | XP_011521756.2 | |
XM_011523458.2 | 2622 | Missense Mutation | AAC,GAC | N,D 588 | XP_011521760.1 | |
XM_017023879.1 | 2622 | Missense Mutation | AAC,GAC | N,D 837 | XP_016879368.1 | |
XM_017023880.1 | 2622 | Missense Mutation | AAC,GAC | N,D 825 | XP_016879369.1 | |
XM_017023881.1 | 2622 | Missense Mutation | AAC,GAC | N,D 818 | XP_016879370.1 | |
XM_017023882.1 | 2622 | Intron | XP_016879371.1 | |||
XM_017023883.1 | 2622 | Intron | XP_016879372.1 | |||
XM_017023884.1 | 2622 | Intron | XP_016879373.1 |