Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAGTGGGAGAATAATTAATGT[G/T]TCTAATGAACTATTACATTTTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300933 | ||||||||||||||||||||
Literature Links: |
NUP62CL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUP62CL - nucleoporin 62 C-terminal like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIH1D3 - PIH1 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169154.1 | Intron | NP_001162625.1 | ||||
NM_173494.1 | Intron | NP_775765.1 | ||||
XM_011530855.2 | Intron | XP_011529157.1 |