Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAACGCTGGTTAATCCCCGGGCA[C/T]TGAGGATACATGGAAGGCTCAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150100 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDHB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDHB - lactate dehydrogenase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174097.2 | 1223 | Missense Mutation | AAT,AGT | N,S 297 | NP_001167568.1 | |
NM_001315537.1 | 1223 | Intron | NP_001302466.1 | |||
NM_002300.7 | 1223 | Intron | NP_002291.1 | |||
XM_006719074.2 | 1223 | Intron | XP_006719137.1 |