Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTTATTTCTTTGCCATACAAAG[C/G]ATAGTAGTCTGGTTTAAGAATCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300628 MIM: 300547 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMD7 - FERM domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306193.1 | 2130 | Missense Mutation | CCT,GCT | P,A 649 | NP_001293122.1 | |
NM_194277.2 | 2130 | Missense Mutation | CCT,GCT | P,A 664 | NP_919253.1 | |
XM_017029947.1 | 2130 | Missense Mutation | CCT,GCT | P,A 648 | XP_016885436.1 | |
XM_017029948.1 | 2130 | Missense Mutation | CCT,GCT | P,A 579 | XP_016885437.1 | |
XM_017029949.1 | 2130 | Missense Mutation | CCT,GCT | P,A 506 | XP_016885438.1 |
STK26 - serine/threonine protein kinase 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |