Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACCCGAAGGTTACCCTGACCAT[C/T]TTTGTTGCCCGCCTCTACTACTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607113 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC3G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOBEC3G - apolipoprotein B mRNA editing enzyme catalytic subunit 3G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021822.3 | 711 | Silent Mutation | ATC,ATT | I,I 118 | NP_068594.1 | |
XM_006724290.1 | 711 | Silent Mutation | ATC,ATT | I,I 51 | XP_006724353.1 | |
XM_017028903.1 | 711 | Silent Mutation | ATC,ATT | I,I 118 | XP_016884392.1 | |
XM_017028904.1 | 711 | Silent Mutation | ATC,ATT | I,I 51 | XP_016884393.1 |