Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATAATGGAGTCTGAGTATGTCCT[A/G]TGCAACTGGAAAGACCAGTTGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUM1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MUM1L1 - MUM1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171020.1 | 664 | Silent Mutation | CTA,CTG | L,L 7 | NP_001164491.1 | |
NM_152423.4 | 664 | Silent Mutation | CTA,CTG | L,L 7 | NP_689636.3 | |
XM_005262077.2 | 664 | Silent Mutation | CTA,CTG | L,L 7 | XP_005262134.1 | |
XM_011530856.1 | 664 | Silent Mutation | CTA,CTG | L,L 7 | XP_011529158.1 | |
XM_011530857.1 | 664 | Silent Mutation | CTA,CTG | L,L 7 | XP_011529159.1 | |
XM_017029269.1 | 664 | Silent Mutation | CTA,CTG | L,L 7 | XP_016884758.1 |