Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGCGGGCCGCGCTCGCGGCGGA[C/G]AACGCGGATGGGGAACCCGGCGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603330 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAH9 - dynein axonemal heavy chain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001372.3 | 71 | Missense Mutation | GAC,GAG | D,E 13 | NP_001363.2 | |
NM_004662.2 | 71 | Intron | NP_004653.2 | |||
XM_011523703.1 | 71 | Missense Mutation | GAC,GAG | D,E 13 | XP_011522005.1 | |
XM_017024292.1 | 71 | Missense Mutation | GAC,GAG | D,E 13 | XP_016879781.1 | |
XM_017024293.1 | 71 | Intron | XP_016879782.1 | |||
XM_017024294.1 | 71 | Missense Mutation | GAC,GAG | D,E 13 | XP_016879783.1 | |
XM_017024295.1 | 71 | Missense Mutation | GAC,GAG | D,E 13 | XP_016879784.1 |