Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601102 MIM: 102577 MIM: 180647 MIM: 611334 | |||||||||||||||||||||||
Literature Links: |
EIF4A2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EIF4A2 - eukaryotic translation initiation factor 4A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001967.3 | 1353 | Intron | NP_001958.2 |
MIR1248 - microRNA 1248 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFC4 - replication factor C subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002916.3 | 1353 | Silent Mutation | CAA,CAG | Q,Q 357 | NP_002907.1 | |
NM_181573.2 | 1353 | Silent Mutation | CAA,CAG | Q,Q 357 | NP_853551.1 |
SNORA4 - small nucleolar RNA, H/ACA box 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA63 - small nucleolar RNA, H/ACA box 63 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA81 - small nucleolar RNA, H/ACA box 81 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD2 - small nucleolar RNA, C/D box 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |