Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCGGGTGCCGCGGCCAGTGGAGC[C/T]GCCAGGGGACCTGGGTCTGCCAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611179 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHROOM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHROOM1 - shroom family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOWAHA - sosondowah ankyrin repeat domain family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175873.5 | 690 | Missense Mutation | CCG,CTG | P,L 131 | NP_787069.4 |