Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAAGCTGGCCACAGAGACAGGTC[A/G]TTCCCATTAGGATTCCTAGAAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CES5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CES5A - carboxylesterase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143685.1 | 1661 | Silent Mutation | AAC,AAT | N,N 504 | NP_001137157.1 | |
NM_001190158.1 | 1661 | Silent Mutation | AAC,AAT | N,N 533 | NP_001177087.1 | |
NM_145024.2 | 1661 | Silent Mutation | AAC,AAT | N,N 454 | NP_659461.1 |