Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCTCCAGGCGCTGCCTCAGCG[A/G]GATGAGCACCAGCTCCACCACGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116949 MIM: 117140 MIM: 610674 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC25B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC25B - cell division cycle 25B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CENPB - centromere protein B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPEF1 - sperm flagellar 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015417.4 | 495 | Missense Mutation | CCG,TCG | P,S 106 | NP_056232.2 | |
XM_005260683.4 | 495 | Missense Mutation | CCG,TCG | P,S 106 | XP_005260740.1 |