Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTAAATAAATGAAAAATGCACC[A/G]AGCCAAACAAAACTTAACTGGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607909 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AZIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AZIN1 - antizyme inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301668.1 | 2415 | UTR 3 | NP_001288597.1 | |||
NM_015878.5 | 2415 | UTR 3 | NP_056962.2 | |||
NM_148174.3 | 2415 | UTR 3 | NP_680479.1 | |||
XM_005250969.1 | 2415 | UTR 3 | XP_005251026.1 | |||
XM_011517126.1 | 2415 | UTR 3 | XP_011515428.1 | |||
XM_011517127.1 | 2415 | UTR 3 | XP_011515429.1 | |||
XM_011517128.2 | 2415 | UTR 3 | XP_011515430.1 | |||
XM_017013563.1 | 2415 | UTR 3 | XP_016869052.1 | |||
XM_017013564.1 | 2415 | UTR 3 | XP_016869053.1 | |||
XM_017013565.1 | 2415 | UTR 3 | XP_016869054.1 | |||
XM_017013566.1 | 2415 | UTR 3 | XP_016869055.1 |
LOC107986873 - forkhead box protein L2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017014121.1 | 2415 | Intron | XP_016869610.1 |