Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGGGAGACAGTGCTCACGAAC[C/T]GGAGAACCCAGGTATTTGGGGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609921 MIM: 616955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP10 - LDL receptor related protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REM2 - RRAD and GEM like GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173527.2 | 1706 | Missense Mutation | CCG,CTG | P,L 145 | NP_775798.2 | |
XM_005267383.3 | 1706 | Missense Mutation | CCG,CTG | P,L 145 | XP_005267440.1 | |
XM_011536518.2 | 1706 | Missense Mutation | CCG,CTG | P,L 177 | XP_011534820.1 | |
XM_017021058.1 | 1706 | Missense Mutation | CCG,CTG | P,L 177 | XP_016876547.1 | |
XM_017021059.1 | 1706 | Missense Mutation | CCG,CTG | P,L 177 | XP_016876548.1 |