Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAGTTCACCAGCACGGACTCC[A/G]TCAGGATGGAGGCCAGGATCTGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf45 - chromosome 5 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100996419 - uncharacterized LOC100996419 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D9B - TBC1 domain family member 9B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015043.3 | 3612 | Missense Mutation | ACG,ATG | T,M 1192 | NP_055858.2 | |
NM_198868.2 | 3612 | Missense Mutation | ACG,ATG | T,M 1209 | NP_942568.2 |