Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGTTTGGATTTAATGCTGCCCA[A/G]GACTTCTGGTGTTCAACTTTGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRB3 - adhesion G protein-coupled receptor B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001704.2 | 679 | Silent Mutation | CAA,CAG | Q,Q 26 | NP_001695.1 | |
XM_005248752.2 | 679 | Silent Mutation | CAA,CAG | Q,Q 26 | XP_005248809.1 | |
XM_006715534.3 | 679 | Silent Mutation | CAA,CAG | Q,Q 26 | XP_006715597.1 | |
XM_011536009.2 | 679 | Intron | XP_011534311.1 | |||
XM_011536010.2 | 679 | UTR 5 | XP_011534312.1 | |||
XM_011536011.2 | 679 | UTR 5 | XP_011534313.1 | |||
XM_011536012.2 | 679 | Silent Mutation | CAA,CAG | Q,Q 26 | XP_011534314.1 | |
XM_017011140.1 | 679 | Silent Mutation | CAA,CAG | Q,Q 26 | XP_016866629.1 | |
XM_017011141.1 | 679 | Intron | XP_016866630.1 | |||
XM_017011142.1 | 679 | Silent Mutation | CAA,CAG | Q,Q 26 | XP_016866631.1 | |
XM_017011143.1 | 679 | Silent Mutation | CAA,CAG | Q,Q 26 | XP_016866632.1 | |
XM_017011144.1 | 679 | Silent Mutation | CAA,CAG | Q,Q 26 | XP_016866633.1 |
LOC101928307 - uncharacterized LOC101928307 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |