Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACATGAACCTCATGGATGCCTCC[A/G]TGCCTCCCCTGGCCCTTGGGCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600659 MIM: 606422 MIM: 613701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
E2F4 - E2F transcription factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELMO3 - engulfment and cell motility 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024712.3 | 755 | Missense Mutation | ATG,GTG | M,V 238 | NP_078988.2 |
LOC105369155 - uncharacterized LOC105369155 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC29 - leucine rich repeat containing 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR328 - microRNA 328 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |