Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAAGGTGGACATCATCATCAGC[A/G]AGTGGATGGGCTACTGCCTCTTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608846 MIM: 602950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADM5 - adrenomedullin 5 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPT1C - carnitine palmitoyltransferase 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5088 - microRNA 5088 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRMT1 - protein arginine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207042.2 | 601 | Missense Mutation | AAG,GAG | K,E 144 | NP_001193971.1 | |
NM_001536.5 | 601 | Missense Mutation | AAG,GAG | K,E 162 | NP_001527.3 | |
NM_198318.4 | 601 | Missense Mutation | AAG,GAG | K,E 144 | NP_938074.2 | |
XM_005258842.1 | 601 | Missense Mutation | AAG,GAG | K,E 133 | XP_005258899.1 | |
XM_017026734.1 | 601 | Missense Mutation | AAG,GAG | K,E 138 | XP_016882223.1 | |
XM_017026735.1 | 601 | Missense Mutation | AAG,GAG | K,E 138 | XP_016882224.1 | |
XM_017026736.1 | 601 | Missense Mutation | AAG,GAG | K,E 133 | XP_016882225.1 |