Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGCCTGCGGAGCCTGTGGGGC[A/T]GGTAAGGGGCAAGAGATTCCAGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147220 | ||||||||||||||||||||
Literature Links: |
IGLC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGLC1 - immunoglobulin lambda constant 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGLL5 - immunoglobulin lambda like polypeptide 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178126.1 | 479 | Missense Mutation | AGG,TGG | R,W 69 | NP_001171597.1 | |
NM_001256296.1 | 479 | Silent Mutation | GCA,GCT | A,A 33 | NP_001243225.1 |
MIR5571 - microRNA 5571 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |