Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCTGGAACCTGGAGAAGAGAGA[C/T]GGTGGCAATCCACTCTGCACTCGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608549 | ||||||||||||||||||||
Literature Links: |
VPS11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VPS11 - VPS11, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290185.1 | 396 | Silent Mutation | GAC,GAT | D,D 112 | NP_001277114.1 | |
NM_021729.5 | 396 | Silent Mutation | GAC,GAT | D,D 122 | NP_068375.3 | |
XM_011542898.1 | 396 | Intron | XP_011541200.1 | |||
XM_011542899.2 | 396 | UTR 5 | XP_011541201.1 |