Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACACAAAGGAAGCGGGAGAGGGC[G/A]GAGCGCCACCAGGAGGCTGCGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602302 MIM: 615791 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HR - hair growth associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005144.4 | 5385 | UTR 3 | NP_005135.2 | |||
NM_018411.4 | 5385 | UTR 3 | NP_060881.2 | |||
XM_005273569.1 | 5385 | UTR 3 | XP_005273626.1 | |||
XM_006716367.1 | 5385 | UTR 3 | XP_006716430.1 |
NUDT18 - nudix hydrolase 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |