Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGCCTTGGACTTGATGCAGTCA[G/T]AATATGTTCCATTTTTATAGTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612213 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BSPH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BSPH1 - binder of sperm protein homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128326.2 | 259 | Missense Mutation | TAT,TCT | Y,S 57 | NP_001121798.1 | |
XM_017026118.1 | 259 | Missense Mutation | TAT,TCT | Y,S 57 | XP_016881607.1 |