Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTCAAGAAGGGTCATCATTATA[C/T]TCTGAAGCAGAATTCTTCAGGTACT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607760 | |||||||||||||||||||||||
Literature Links: |
TOPBP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TOPBP1 - topoisomerase (DNA) II binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007027.3 | 4345 | UTR 3 | NP_008958.2 | |||
XM_005247076.3 | 4345 | UTR 3 | XP_005247133.1 | |||
XM_011512357.2 | 4345 | Intron | XP_011510659.1 | |||
XM_017005636.1 | 4345 | UTR 3 | XP_016861125.1 | |||
XM_017005637.1 | 4345 | UTR 3 | XP_016861126.1 |