Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGCTGGGCCCTCAGGAGCCGAC[C/T]GGAAGCCAGTTGCTACGCAGCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609967 MIM: 604951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM129C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM129C - family with sequence similarity 129 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098524.1 | 240 | Silent Mutation | ACC,ACT | T,T 85 | NP_001091994.1 | |
NM_001321826.1 | 240 | Silent Mutation | ACC,ACT | T,T 54 | NP_001308755.1 | |
NM_001321827.1 | 240 | Silent Mutation | ACC,ACT | T,T 54 | NP_001308756.1 | |
NM_001321828.1 | 240 | Intron | NP_001308757.1 | |||
NM_173544.4 | 240 | Silent Mutation | ACC,ACT | T,T 85 | NP_775815.2 | |
XM_005259813.3 | 240 | Silent Mutation | ACC,ACT | T,T 54 | XP_005259870.1 | |
XM_005259814.3 | 240 | Silent Mutation | ACC,ACT | T,T 54 | XP_005259871.1 | |
XM_011527781.2 | 240 | Silent Mutation | ACC,ACT | T,T 54 | XP_011526083.1 | |
XM_011527786.2 | 240 | Silent Mutation | ACC,ACT | T,T 54 | XP_011526088.1 | |
XM_011527787.2 | 240 | Silent Mutation | ACC,ACT | T,T 54 | XP_011526089.1 | |
XM_011527789.1 | 240 | Silent Mutation | ACC,ACT | T,T 54 | XP_011526091.1 | |
XM_017026453.1 | 240 | Silent Mutation | ACC,ACT | T,T 54 | XP_016881942.1 | |
XM_017026454.1 | 240 | Silent Mutation | ACC,ACT | T,T 54 | XP_016881943.1 | |
XM_017026455.1 | 240 | Intron | XP_016881944.1 | |||
XM_017026456.1 | 240 | Intron | XP_016881945.1 | |||
XM_017026457.1 | 240 | Intron | XP_016881946.1 |
PGLS - 6-phosphogluconolactonase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |