Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGGCTTGGGCGCGCGGGACCTG[A/G]CCCAGGACGCTCAGATCACCAGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615776 | ||||||||||||||||||||
Literature Links: |
CROCC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CROCC - ciliary rootlet coiled-coil, rootletin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014675.4 | 161 | Missense Mutation | ACC,GCC | T,A 39 | NP_055490.4 | |
XM_006711058.3 | 161 | Intron | XP_006711121.1 | |||
XM_011542468.1 | 161 | Missense Mutation | ACC,GCC | T,A 41 | XP_011540770.1 | |
XM_011542470.2 | 161 | Missense Mutation | ACC,GCC | T,A 41 | XP_011540772.1 | |
XM_017002913.1 | 161 | Missense Mutation | ACC,GCC | T,A 41 | XP_016858402.1 | |
XM_017002914.1 | 161 | Missense Mutation | ACC,GCC | T,A 41 | XP_016858403.1 | |
XM_017002915.1 | 161 | Missense Mutation | ACC,GCC | T,A 41 | XP_016858404.1 |