Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTGTTCCATGGGTCCTGTCAT[G/T]GGAGAGGACCAAACATCCATCCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611359 MIM: 615086 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMBRA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMBRA1 - autophagy and beclin 1 regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HARBI1 - harbinger transposase derived 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173811.3 | 1543 | Silent Mutation | NP_776172.1 | |||
XM_011520025.1 | 1543 | Silent Mutation | XP_011518327.1 | |||
XM_011520026.2 | 1543 | Silent Mutation | XP_011518328.1 | |||
XM_011520027.1 | 1543 | Silent Mutation | XP_011518329.1 | |||
XM_011520028.2 | 1543 | Silent Mutation | XP_011518330.1 | |||
XM_011520029.2 | 1543 | Silent Mutation | XP_011518331.1 | |||
XM_017017602.1 | 1543 | Silent Mutation | XP_016873091.1 | |||
XM_017017603.1 | 1543 | Silent Mutation | XP_016873092.1 | |||
XM_017017604.1 | 1543 | Silent Mutation | XP_016873093.1 | |||
XM_017017605.1 | 1543 | Silent Mutation | XP_016873094.1 | |||
XM_017017606.1 | 1543 | Silent Mutation | XP_016873095.1 |