Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGGCCGGGCCGACGGAGAGGG[A/C]GAGGGGGAGGAAGAGGAGGGGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607548 MIM: 602388 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSPH6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RSPH6A - radial spoke head 6 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030785.3 | 3717 | Intron | NP_110412.1 | |||
XM_011527351.1 | 3717 | Intron | XP_011525653.1 |
SYMPK - symplekin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004819.2 | 3717 | Silent Mutation | TCG,TCT | S,S 1173 | NP_004810.2 | |
XM_005259286.1 | 3717 | Silent Mutation | TCG,TCT | S,S 1173 | XP_005259343.1 | |
XM_011527354.1 | 3717 | Silent Mutation | TCG,TCT | S,S 1173 | XP_011525656.1 |