Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGCCTTCTAAGCGGCGGGCTAG[G/T]TCCCTGGTAAAAAGAATGTTAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610526 MIM: 616796 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NT5C1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NT5C1B - 5'-nucleotidase, cytosolic IB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NT5C1B-RDH14 - NT5C1B-RDH14 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199103.1 | 811 | Missense Mutation | GAA,GAC | E,D 544 | NP_001186032.1 | |
NM_001199104.1 | 811 | UTR 3 | NP_001186033.1 |
RDH14 - retinol dehydrogenase 14 (all-trans/9-cis/11-cis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020905.3 | 811 | Missense Mutation | GAA,GAC | E,D 230 | NP_065956.1 |