Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATATGGGCAGGAAGAAAGAGACC[C/T]GACAGGCTGAGCACAACCTTCCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603501 | ||||||||||||||||||||
Literature Links: |
PARG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PARG - poly(ADP-ribose) glycohydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303486.1 | 3548 | UTR 3 | NP_001290415.1 | |||
NM_001303487.1 | 3548 | UTR 3 | NP_001290416.1 | |||
NM_001303489.1 | 3548 | UTR 3 | NP_001290418.1 | |||
NM_001324381.1 | 3548 | UTR 3 | NP_001311310.1 | |||
NM_003631.3 | 3548 | UTR 3 | NP_003622.2 | |||
XM_011540305.1 | 3548 | UTR 3 | XP_011538607.1 | |||
XM_011540308.2 | 3548 | Intron | XP_011538610.1 |