Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTCACATTTTACTATCTCTAT[C/G]TCTTCCTGGCTCAGCACCCTTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARD17 - caspase recruitment domain family member 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007232.1 | 134 | Missense Mutation | GAC,GAG | D,E 39 | NP_001007233.1 | |
XM_011542838.2 | 134 | Missense Mutation | GAC,GAG | D,E 54 | XP_011541140.1 |