Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGGGGGGCCTTTAGCACTCAC[A/T]GTCTGTTGTACATTCTTTGTGGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602984 | ||||||||||||||||||||
Literature Links: |
MED6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED6 - mediator complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284209.1 | 749 | Silent Mutation | ACA,ACT | T,T 239 | NP_001271138.1 | |
NM_001284210.1 | 749 | UTR 3 | NP_001271139.1 | |||
NM_001284211.1 | 749 | Silent Mutation | ACA,ACT | T,T 232 | NP_001271140.1 | |
NM_005466.3 | 749 | Silent Mutation | ACA,ACT | T,T 232 | NP_005457.2 |