Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGACTCCAGAGAGCAGCGCTG[A/G]GTGAAGGCTTTATTGCAGACGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616441 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OVOL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OVOL2 - ovo like zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303461.1 | 803 | Silent Mutation | ACC,ACT | T,T 53 | NP_001290390.1 | |
NM_001303462.1 | 803 | Silent Mutation | ACC,ACT | T,T 53 | NP_001290391.1 | |
NM_021220.3 | 803 | Silent Mutation | ACC,ACT | T,T 185 | NP_067043.2 |