Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCACTTGTTTTTTCCTATTAAC[G/T]TGTAAGCGGCGATCAACCAGATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL8 - methyltransferase like 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321154.1 | 984 | Missense Mutation | CAA,CAC | Q,H 368 | NP_001308083.1 | |
NM_001321155.1 | 984 | Missense Mutation | CAA,CAC | Q,H 368 | NP_001308084.1 | |
NM_001321156.1 | 984 | Missense Mutation | CAA,CAC | Q,H 368 | NP_001308085.1 | |
NM_001321157.1 | 984 | Missense Mutation | CAA,CAC | Q,H 416 | NP_001308086.1 | |
NM_001321158.1 | 984 | Missense Mutation | CAA,CAC | Q,H 323 | NP_001308087.1 | |
NM_001321159.1 | 984 | Missense Mutation | CAA,CAC | Q,H 318 | NP_001308088.1 | |
NM_001321160.1 | 984 | UTR 3 | NP_001308089.1 | |||
NM_001321161.1 | 984 | UTR 3 | NP_001308090.1 | |||
NM_001321162.1 | 984 | UTR 3 | NP_001308091.1 | |||
NM_024770.4 | 984 | Missense Mutation | CAA,CAC | Q,H 368 | NP_079046.2 | |
XM_011511875.2 | 984 | Missense Mutation | CAA,CAC | Q,H 353 | XP_011510177.1 | |
XM_017004984.1 | 984 | Missense Mutation | CAA,CAC | Q,H 429 | XP_016860473.1 | |
XM_017004985.1 | 984 | Missense Mutation | CAA,CAC | Q,H 381 | XP_016860474.1 | |
XM_017004986.1 | 984 | Missense Mutation | CAA,CAC | Q,H 381 | XP_016860475.1 | |
XM_017004987.1 | 984 | Missense Mutation | CAA,CAC | Q,H 336 | XP_016860476.1 | |
XM_017004988.1 | 984 | Missense Mutation | CAA,CAC | Q,H 331 | XP_016860477.1 | |
XM_017004989.1 | 984 | Missense Mutation | CAA,CAC | Q,H 381 | XP_016860478.1 |