Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCGGCAGTGGTGCTTGGAGAGC[C/T]TGATGCTGAACTCCTTCCGGCAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616372 MIM: 605471 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF4 - DDB1 and CUL4 associated factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFYVE1 - zinc finger FYVE-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281734.1 | 1049 | Missense Mutation | AAG,AGG | K,R 717 | NP_001268663.1 | |
NM_001281735.1 | 1049 | Missense Mutation | AAG,AGG | K,R 316 | NP_001268664.1 | |
NM_021260.3 | 1049 | Missense Mutation | AAG,AGG | K,R 731 | NP_067083.1 | |
NM_178441.2 | 1049 | Missense Mutation | AAG,AGG | K,R 316 | NP_848535.1 | |
XM_017021373.1 | 1049 | Missense Mutation | AAG,AGG | K,R 316 | XP_016876862.1 |