Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTGCTGTCTGTCCCAGGGATGG[A/G]GAGGACTGCTCCAGGGAAGCTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA2R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANXA2R - annexin A2 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014279.2 | 679 | Missense Mutation | CCC,CTC | P,L 127 | NP_001014301.1 |
LOC153684 - uncharacterized LOC153684 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC648987 - uncharacterized LOC648987 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |