Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATCGCAGGCCCAGACGGCAGAA[A/G]CTTCAGTGGTCAGCAGCCCATAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606903 MIM: 600560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928120 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928120 - uncharacterized LOC101928120 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PBXIP1 - PBX homeobox interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYGO2 - pygopus family PHD finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138300.3 | 1324 | Missense Mutation | GCT,GTT | A,V 373 | NP_612157.1 |
SHC1 - SHC adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |