Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATCGCAAAATCCAGGCCTCAGA[A/G]GCCTCCGAGACCCCTATGGCCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGEH1 - MAGE family member H1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014061.4 | 373 | Silent Mutation | GAA,GAG | E,E 29 | NP_054780.2 |
MIR4536-1 - microRNA 4536-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4536-2 - microRNA 4536-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |